Isotemp™ Model 282A Vacuum Oven
Isotemp™ Model 282A Vacuum Oven
Actual product may vary
Isotemp™ Model 282A Vacuum Oven
Ideal for general vacuum drying and heating applications.
Have Questions?
Catalog NumberVoltageWattage
FIS13-262-52120 V1800 w
Catalog number FIS13-262-52
Price (SGD)
-
Voltage:
120 V
Wattage:
1800 w
Ideal for general vacuum drying and heating applications.
Designed to operate with reduced pressures and/or inert atmospheres, oven is equipped with separate vacuum and gas ports, each with its own control valve.
Operating Features
  • Capacity: 1.5 cu. ft. (0.04m3)
  • Temperature range: Ambient to 280°C (536°F)
  • Automatic time and temperature control withe LED display
  • Seven-day programmable timer can cycle the oven on or off up to eight times per day
  • Self diagnostics for open thermocouple, shorted triac, open heater, and overtemperature prevention plus panel alert lights
  • Built-in safety controller prevents temperature runaway
  • Backup thermostat
Cabinet Construction
  • Chamber door with full-view, tempered safety glass
  • Silicone door gasket and stainless-steel interior
  • Cold-rolled steel exterior is protected by baked-on epoxy enamel finish
  • Can hold up to eight shelves
When vacuum and/or inert gases are used, opening and closing purge and vacuum valves are also required.
Specifications
Capacity (English)1.5 cu.ft.
Chamber MaterialSteel
Capacity (Metric)42.4753 L
Temperature (Metric)280°C
TypeVacuum Oven
DisplayLED
IncludesTwo shelves and a power cord with plug
MaterialBaked Epoxy Enamel Coated Stainless Steel
No. of Shelves2
PortsVacuum and gas
Safety FeaturesElectronic control back up prevents overheating
Temperature Resolution1°C
Temperature Uniformity±5° to 100°C
Vacuum Range0 to 30 in.Hg
Length (English)23 in.
Length (Metric)28 cm
Width (English)26 in.
Width (English) Interior12 in.
Height (English)22 in.
Height (English) Interior12 in.
Height (Metric)56 cm
Height (Metric) Interior31 cm
Depth (English) Interior18 in.
Depth (Metric) Interior46 cm
Shipping Weight (English)205 lb.
Electrical Requirements120 V 50/60 Hz
Voltage120 V
Wattage1800 w
Amperage14.7 A