Kimble™ KIMAX™ Babcock Bottles for Cream Testing
Kimble™ KIMAX™ Babcock Bottles for Cream Testing
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Babcock Bottles for Cream Testing
With Sealed 3 marking
Have Questions?
Catalog NumberCapacity (Metric)Tolerance
FIS12-700-3010 mL±0.5%
Catalog number FIS12-700-30
Price (SGD)
959.00
CS
Add to cart
Capacity (Metric):
10 mL
Tolerance:
±0.5%
Price (SGD)
959.00
CS
Add to cart
With Sealed 3 marking
  • Bottles graduated to 50% in 0.5% subdivisions Graduations are marked with permanent brown stain Overall height: 165mm Marked “Sealed 3” for states requiring this special designation
Specifications
Capacity0.33 oz., 10 mL
Capacity (Metric)10 mL
Capacity (English)0.33 oz.
MaterialGlass
Tolerance±0.5%
Height (Metric)165 mm
Height (English)6.5 in.
Unit SizeCS