Nunc™ IVF Multi Dish, 4-well
Actual product may vary
Thermo Scientific
Nunc™ IVF Multi Dish, 4-well
Perform applications which yield consistently reliable, high-quality results during vitrification.
Have Questions?
Catalog NumberSurface Treatment
FIS12-566-350Non-treated
Catalog number FIS12-566-350
Price (SGD)
1,929.00
Case
Surface Treatment:
Non-treated
Price (SGD)
1,929.00
Case
Generate consistently reliable results during IVF and embryo vitrification with Thermo Scientific™ Nunc™ 4-Well IVF Dishes, which meet the rigorous requirements of IVF testing, certification and validation.
Innovative features for high quality and ease-of-use:
  • Ensures proper well orientation and prevents cross contamination with notched lid and dish
  • Prevents scratches on well bottoms and promotes fast temperature equilibrium with 0.8mm raised edge
  • The Nunclon Delta surface treatment ensures consistency of a more hydrophilic surface
  • The non-treated hydrophobic surface facilitates microdrop embryo culture
  • Certificates of conformity available for every lot
Rigorously tested, certified and validated for IVF:
  • CE-marked (Notified Body 2460), FDA-listed
  • Produced under ISO 13485 with full lot traceability
  • Sterile (SAL 10-6)
  • Non-pyrogenic (LAL tested)
  • USP Class VI polystyrene
  • Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested
DescriptionNon-treated IVF Dish, 4-well
Culture Area1.9 cm2/well
MaterialPolystyrene (USP Class VI)
SterilitySterile
Surface TreatmentNon-treated
Well Count4
No. per Case120
No. per Pack4
Length (Metric)66 mm
Width (Metric)66 mm
Unit SizeCase