Nunc™ Cell Scrapers
Actual product may vary
Thermo Scientific
Nunc™ Cell Scrapers
Conduct a variety of applications with sterile, nonpyrogenic Thermo Scientific™ Nunc™ Cell Scrapers, which feature an adjustable blade.
Have Questions?
Catalog NumberLength (Metric) Handle
FIS12-565-5932 cm
Catalog number FIS12-565-59
Price (SGD)
795.00
Pack
Length (Metric) Handle:
32 cm
Price (SGD)
795.00
Pack
Featuring an adjustable blade, the sterile, nonpyrogenic Thermo Scientific™ Nunc™ Cell Scraper allows for convenient mechanical harvesting of the adherent cells from culture vessels. Comes in 2 lengths to accommodate wide range of culture wares.
Features:
  • Flexible polystyrene handles
  • Sterilized and individually wrapped
Ordering Information:
  • 179707: for use with 75 to 175cm2 flasks
  • 179693: for use with 25 to 80cm2 flasks
Warranty: 90 days.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Length (English) Blade0.6299 in.
Length (Metric) Blade17.5 mm
Width (English) Blade0.6889 in.
Width (Metric) Blade17.5 mm
For Use With (Equipment)75 to 175 cm&Sup2; Flask
Length (English) Handle12.5 in.
Length (Metric) Handle32 cm
No. per Pack1/Pk.
No. per Case250/Cs.
Product TypeCell Scraper
SterilitySterile
MaterialPolystyrene (Handle)
Unit SizePack