Search Thermo Fisher Scientific
| Catalog Number | Holds |
|---|---|
| FIS12-565-178 | 40 Cryogenic Tubes |
| 5030-0510 | 50 Cryogenic Tubes |
| FIS03-337-7E | 50 Cryogenic Tubes |
| 5030-0505 | 25 Cryogenic Tubes |
| FIS03-337-7Z | 25 Cryogenic Tubes |
Expand the functionality of cryogenic tube products with Thermo Scientific™ CryoTube™ Holders — benchtop tube and vial containers for single-handed capping and decapping. The product range includes Nalgene™ and Nunc™ containers in various sizes and compartment configurations.