Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Actual product may vary
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Thermo Scientific
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Store samples from general cold storage to the vapor phase of liquid nitrogen with these biobanking and cell culture cryogenic tubes, available with either internal or external threads.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBottom ShapeCapacity (Metric)Thread StyleStyle
FIS12-565-171NRound (Self-standing)1.8 mLInternalWriting Surface
374080Conical (Self-standing)0.5 mLInternalNo Writing Surface, A-R Base
375353Conical (Self-standing)1 mLExternalWriting Surface, A-R Base
FIS12-565-164NConical (Self-standing)1 mLExternalWriting Surface, A-R Base
FKS10544641Conical (Self-standing)1 mLExternalWriting Surface, A-R Base
NNC375353Conical (Self-standing)1 mLExternalWriting Surface, A-R Base
FIS12-565-297Conical (Self-standing)1 mLExternalNo Writing Surface, A-R Base
NNC377224Conical (Self-standing)1 mLInternalWriting Surface, A-R Base
366656Conical (Self-standing)1 mLInternalWriting Surface
375418Round (Self-standing)1.8 mLExternalWriting Surface, A-R Base
NNC375418Round (Self-standing)1.8 mLExternalWriting Surface, A-R Base
347627Round (Self-standing)1.8 mLExternalNo Writing Surface, A-R Base
FKS10080601Round (Self-standing)1.8 mLExternalNo Writing Surface, A-R Base
NNC347627Round (Self-standing)1.8 mLExternalNo Writing Surface, A-R Base
377267Round (Self-standing)1.8 mLInternalWriting Surface, A-R Base
FIS12-565-167NRound (Self-standing)1.8 mLInternalWriting Surface, A-R Base
FKS11341675Round (Self-standing)1.8 mLInternalWriting Surface, A-R Base
368632Round (Self-standing)1.8 mLInternalWriting Surface
FKS10674511Round (Self-standing)1.8 mLInternalWriting Surface
NNC368632Round (Self-standing)1.8 mLInternalWriting Surface
363401Round1.8 mLInternalWriting Surface
FKS10004220Round1.8 mLInternalWriting Surface
NNC363401Round1.8 mLInternalWriting Surface
351934Round1.8 mLInternalWriting Surface, No Cap
379189Round (Self-standing)3.6 mLInternalWriting Surface, A-R Base
FIS12-565-162NRound (Self-standing)3.6 mLInternalWriting Surface, A-R Base
NNC379189Round (Self-standing)3.6 mLInternalWriting Surface, A-R Base
FKS10789871Round3.6 mLInternalWriting Surface
337516Round (Self-standing)4.5 mLExternalWriting Surface, A-R Base
FIS12-565-291Round (Self-standing)4.5 mLExternalWriting Surface, A-R Base
NNC337516Round (Self-standing)4.5 mLExternalWriting Surface, A-R Base
347643Round (Self-standing)4.5 mLExternalNo Writing Surface, A-R Base
FIS12-565-161NConical (Self-standing)4.5 mLInternalWriting Surface, A-R Base
FKS10716922Conical (Self-standing)4.5 mLInternalWriting Surface, A-R Base
363452Round4.5 mLInternalWriting Surface
FIS12-565-173NRound4.5 mLInternalWriting Surface
FKS10358131Round4.5 mLInternalWriting Surface
Catalog number FIS12-565-171N
Price (SGD)
1,443.00
Pack
Bottom Shape:
Round (Self-standing)
Capacity (Metric):
1.8 mL
Thread Style:
Internal
Style:
Writing Surface
Price (SGD)
1,443.00
Pack
Store samples from general cold storage to the vapor phase of liquid nitrogen with Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes, available with either internal or external threads.
Features:
  • Non-cytotoxic, non-mutagenic raw materials and pyrogen-free IVD compatible tubes for added sample security
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • Bulk, uncapped, non-sterile versions available for less demanding applications
Specifications
Diameter (Metric)12.5 mm
Product TypeCryogenic Storage Tube
Diameter (English) Outer0.492 in.
Capacity (Metric)1.8 mL
Bottom ShapeRound (Self-standing)
MaterialPolypropylene
Thread StyleInternal
Length (English)1.929 in.
Length (Metric)49 mm
No. per Pack450
StyleWriting Surface
RNase FreeYes
Vapor LN2 (to -150°C)Yes
Product LineNunc
SterilitySterile
Anti-RotationNo
Packaging Quantity450 Pk.
Closure MaterialPolypropylene
Capacity (English)0.06086 oz.
Dnase FreeYes
Closure TypeScrew Cap
PrintedYes
Unit SizePack