Premium Frosted Microscope Slides, Superfrost marking area
Premium Frosted Microscope Slides, Superfrost marking area
Actual product may vary
Premium Frosted Microscope Slides, Superfrost marking area
With Superfrost™ marking area on one end, one side
Have Questions?
Catalog NumberDimensions (L x W)
FIS12-544-275 x 25 mm
Catalog number FIS12-544-2
Price (SGD)
110.00
Pack
Add to cart
Dimensions (L x W):
75 x 25 mm
Price (SGD)
110.00
Pack
Add to cart
With Superfrost™ marking area on one end, one side
  • Excellent retention of markings in all normal procedures
Specifications
FrostedSuperFrost Marking Area 1 End, 1 Side
Length (Metric)75 mm
TypeMicroscope Slide
Width (Metric)25 mm
Length (English)3 in.
Width (English)1 in.
Dimensions (L x W)75 x 25 mm
Unit SizePack