Gold Seal™ Cover Glasses, rectange, 40 x 22mm, Thickness 1
Actual product may vary
Gold Seal™ Cover Glasses, rectange, 40 x 22mm, Thickness 1
Gold Seal™ Cover Glasses, rectange, 40 x 22mm, Thickness 1
Gold Seal™ Cover Glasses, rectange, 40 x 22mm, Thickness 1
Gold Seal™ Cover Glasses, rectange, 40 x 22mm, Thickness 1
Thermo Scientific
Gold Seal™ Cover Glasses, rectange, 40 x 22mm, Thickness 1
Protect and preserve specimens on slides with Epredia™ Gold Seal™ Cover Glasses, made of noncorrosive borosilicate glass.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThickness (Metric)Dimensions (L x W)Shape
FIS12-518-105K#1 (0.13 to 0.16 mm)22 x 22 mmSquare
FIS12-518-105E#1 (0.13 to 0.16 mm)50 x 24 mmRectangle
Catalog number FIS12-518-105K
Price (SGD)
-
Thickness (Metric):
#1 (0.13 to 0.16 mm)
Dimensions (L x W):
22 x 22 mm
Shape:
Square
Each case of 10 ounces converts into a convenient 100-slide storage file with a protective clear cover. Applications include hematology, histology, cytology, microbiology, and urology. Available in a variety of thicknesses and sizes.
  • Virtually colorless appearance
  • Very low levels of inclusions, striae, bubbles, streaks
  • High special transmission
  • Finely adaptive refractive index suited for microscopes
  • Low auto-fluorescence and high chemical resistance
  • Made of noncorrosive borosilicate glass
  • Sealed in a moisture-resistant plastic case with desiccant packs
Specifications
ShapeSquare
MaterialBorosilicate Glass
Length (Metric)22 mm
Width (Metric)22 mm
Thickness (Metric)#1 (0.13 to 0.16 mm)
Dimensions (L x W)22 x 22 mm
QuantityApprox. 162/bx., 1620/cs.
Unit SizePack