▸
Surgical and Dissection Equipment
▸
Surgical and Dissection Forceps and Tweezers
▸
Mid Grade Babcock Tissue Forceps
Actual product may vary
Integra Miltex
Mid Grade Babcock Tissue Forceps
8mm jaw width
Have Questions?
Catalog Number
Length (Metric)
FIS12-460-761
159 mm
Catalog number
FIS12-460-761
Price (SGD)
106.00
Each
Add to cart
Length (Metric):
159 mm
Price (SGD)
106.00
Each
Add to cart
Stainless steel construction
Specifications
Product Type
Forceps, Tissue, Babcock
Material
Stainless Steel
Grade
Mid
Quantity
Each
Length (English)
6.25 in.
Length (Metric)
159 mm