▸
Surgical and Dissection Equipment
▸
Precision Handling Forceps and Tweezers
▸
Foerster Sponge Forcep
Actual product may vary
Integra Miltex
Foerster Sponge Forcep
Satin-finish stainless steel
Have Questions?
Catalog Number
Tip Style
FIS12-460-564
Straight, Serrated
Catalog number
FIS12-460-564
Price (SGD)
311.00
Each
Add to cart
Tip Style:
Straight, Serrated
Price (SGD)
311.00
Each
Add to cart
Straight jaws
Pattern length 9.5 in. (24.1cm)
Specifications
Tip Style
Straight, Serrated
Length (English)
9.5 in.
Material
Stainless Steel
Length (Metric)
241 mm
Product Type
Forceps, Sponge, Foerster
Quantity (No. per Case)
Each
Finish
Satin
Grade
Standard