Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Surgical and Dissection Equipment
▸
Scalpels
▸
Sterile Safety Scalpels
Actual product may vary
Integra Miltex
Sterile Safety Scalpels
Stainless steel, razor-sharp cutting edge for superior cutting and clean incisions
Have Questions?
Catalog Number
Size
Includes
FIS12-460-459
# 11
–
Catalog number
FIS12-460-459
Price (SGD)
119.00
Pack
Add to cart
Size:
# 11
Includes:
–
Price (SGD)
119.00
Pack
Add to cart
Sterile
Stainless-steel, razor-sharp cutting edge
Metric ruler on each scalpel
Individually packaged
Specifications
Blade Material
Stainless Steel
Product Type
Safety Scalpel
Size
# 11
Disposable
Single-use
Sterility
Sterile
Includes
–
Certifications/Compliance
OSHA
Handle Material
Plastic
Shelf Life
5 Years