Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Cleaners and Disinfectants
▸
Labware Detergents and Cleaners
▸
Surgical Instrument Cleaner
Actual product may vary
Integra Miltex
Surgical Instrument Cleaner
Neutral pH (7) detergent
Have Questions?
Catalog Number
Volume (English)
FIS12-460-423
8 oz.
Catalog number
FIS12-460-423
Price (SGD)
496.00
Case
Add to cart
Volume (English):
8 oz.
Price (SGD)
496.00
Case
Add to cart
To remove organic material from instruments
Biodegradable
Rinses-free, no residue
Concentrated
Phosphate-free
Specifications
Form
Liquid
pH
7
Container Type
Bottle
For Use With (Application)
To remove organic material from instruments
Volume (English)
8 oz.