Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Surgical and Dissection Equipment
▸
Biopsy Punches
▸
Miltex™ Standard Biopsy Punches
Actual product may vary
Integra
Miltex™ Standard Biopsy Punches
Stainless-steel razor sharp cutting edge
Have Questions?
Change view
Catalog Number
Diameter (Metric)
FIS12-460-414
0.2, 0.3, 0.4, 0.5, 0.6 cm
FIS12-460-399
0.2 cm
FIS12-460-400
0.15 cm
FIS12-460-404
0.25 cm
FIS12-460-408
0.35 cm
FIS12-460-409
0.4 cm
FIS12-460-412
0.6 cm
FIS12-460-413
0.8 cm
8 Options
Catalog number
FIS12-460-414
Price (SGD)
476.00
Pack
Add to cart
Diameter (Metric):
0.2, 0.3, 0.4, 0.5, 0.6 cm
Price (SGD)
476.00
Pack
Add to cart
Stainless-steel razor sharp cutting edge
Sealed in a sterile, peel-apart, see-through package
Sterilized by ethylene oxide
Single use
Specifications
Blade Material
Stainless Steel
Product Type
Biopsy Punch
For Use With (Application)
Offer the Convenience of Single-use Disposability without Sacrificing quality, Feel or Performance
Handle Type
Ribbed
Description
Seamless
Disposable
Single-use
Sterility
Sterile
Quantity
50/Pk.
Shelf Life
5 Years
Diameter (English)
0.08, 0.12, 0.16, 0.20, 0.24 in.
Diameter (Metric)
0.2, 0.3, 0.4, 0.5, 0.6 cm