Miltex™ Standard Biopsy Punches
Miltex™ Standard Biopsy Punches
Actual product may vary
Integra
Miltex™ Standard Biopsy Punches
Stainless-steel razor sharp cutting edge
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FIS12-460-4120.6 cm
FIS12-460-3990.2 cm
FIS12-460-4000.15 cm
FIS12-460-4040.25 cm
FIS12-460-4080.35 cm
FIS12-460-4090.4 cm
FIS12-460-4140.2, 0.3, 0.4, 0.5, 0.6 cm
FIS12-460-4110.5 cm
FIS12-460-4130.8 cm
Catalog number FIS12-460-412
Price (SGD)
528.00
PK
Add to cart
Diameter (Metric):
0.6 cm
Price (SGD)
528.00
PK
Add to cart
Stainless-steel razor sharp cutting edge
  • Sealed in a sterile, peel-apart, see-through package
  • Sterilized by ethylene oxide
  • Single use
Specifications
Blade MaterialStainless Steel
Product TypeBiopsy Punch
For Use With (Application)Offer the Convenience of Single-use Disposability without Sacrificing quality, Feel or Performance
Handle TypeRibbed
Length (English)2.75 in.
Length (Metric)7 cm
DescriptionSeamless
DisposableSingle-use
SterilitySterile
Quantity50/Pk.
Shelf Life5 Years
Diameter (English)0.23 in.
Diameter (Metric)0.6 cm
Width (English)2 in.
Width (Metric)5 cm
Unit SizePK