Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Specialty Lab Equipment, Instruments, and Apparatuses
▸
General Purpose Forceps and Tweezers
▸
Miltex™ Swiss Jeweler-Style Forceps
Actual product may vary
Integra
Miltex™ Swiss Jeweler-Style Forceps
Have Questions?
Catalog Number
Length (Metric)
Tip Style
Style
FIS12-460-243
120 mm
Narrow, Fine
3
Catalog number
FIS12-460-243
Price (SGD)
196.00
Each
Add to cart
Length (Metric):
120 mm
Tip Style:
Narrow, Fine
Style:
3
Price (SGD)
196.00
Each
Add to cart
Stainless steel
Non-magnetic
Microfine, pointed tips
Specifications
Length (English)
4.75 in.
Length (Metric)
120 mm
Material
Stainless Steel
Quantity
Each
Style
3
Tip Style
Narrow, Fine
Product Type
Forceps, Swiss Micro Jeweler