Depyrogenated Sterile Empty Vials
Depyrogenated Sterile Empty Vials
Actual product may vary
Depyrogenated Sterile Empty Vials
Depyrogenated Sterile Empty Vials
Depyrogenated Sterile Empty Vials
Depyrogenated Sterile Empty Vials
Depyrogenated Sterile Empty Vials
Depyrogenated Sterile Empty Vials
Thermo Scientific
Depyrogenated Sterile Empty Vials
Thermo Scientific™ Sterile Empty Vials are offered in capacities ranging from 1 to 100mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ColorClosure SizeNo. per Case
FIS12-200-132100 mLAmber20 mm50
FIS12-200-1535 mLClear20 mm50
FIS500572835010 mLClear20 mm50
FIS12-200-14610 mLAmber20 mm50
FIS12-200-13430 mLAmber20 mm50
FIS12-200-13550 mLAmber20 mm50
Catalog number FIS12-200-132
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
100 mL
Color:
Amber
Closure Size:
20 mm
No. per Case:
50
Thermo Scientific™ Depyrogenated Sterile Empty Vials are offered in capacities ranging from 1 to 100mL. Certificate of Sterility & Pyrogen Test included with each lot. Sterile vials are Type I borosilicate, assembled with butyl stoppers and aluminum seals.
  • Type 1 Borosilicate glass
  • Assembled wiht butyl stoppers and aluminum seals
  • Suitable for uses where aseptic protocol is required
  • Certificate of Sterility & Pyrogen Test included with each lot
Specifications
DescriptionAmber glass vial pre-assembled with stopper and aluminum crimp seal
Diameter (English) Outer1.75 in.
ColorAmber
Diameter (Metric) Outer44.45 mm
MaterialGlass
SterilitySterile
Volume (Metric)100 mL
Height (English)3.75 in.
No. per Case50
Volume (English)3.4 oz.
Volume (Metric) Total100 mL
Closure MaterialAluminum
Closure TypeCrimp Cap (Assembled) with Stopper
Height (Metric)95.25 mm
Quantity50 Cs.
Certifications/ComplianceCertificates of Sterility and Pyrogen Test
TypeVial
Closure Size20 mm
For Use With (Equipment)Aseptic Protocol
Unit SizeCS