RNase-Free Disposable Pellet Pestles
Actual product may vary
RNase-Free Disposable Pellet Pestles
RNase-Free Disposable Pellet Pestles
RNase-Free Disposable Pellet Pestles
Individually wrapped to ensure purity
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesCapacity (English)
FIS12-141-368With tube0.05 oz.
FIS12-141-364Without tube0.05 oz.
FKS13236679Without tube0.05 oz.
Catalog number FIS12-141-368
Price (SGD)
184.00
Case
Includes:
With tube
Capacity (English):
0.05 oz.
Price (SGD)
184.00
Case
Individually wrapped to ensure purity
  • RNase, DNase and Pyrogen-free
  • Volume: 1.5mL
Specifications
AutoclavableAutoclavable
ColorBlue
Capacity (English)0.05 oz.
Capacity (Metric)1.5 mL
MaterialPolypropylene
Product TypePellet Pestle
DisposableSingle-use
Length (English)2.75 in.
Dnase FreeYes
IncludesWith tube
Length (Metric)70mm
Capacity0.05 oz., 1.5mL
Unit SizeCase