Hastings Triplet Adjustable Focus Measuring Magnifying Glass: Scale
Hastings Triplet Adjustable Focus Measuring Magnifying Glass: Scale
Actual product may vary
Bausch and Lomb
Hastings Triplet Adjustable Focus Measuring Magnifying Glass: Scale
For use with magnifying glasses
Have Questions?
Catalog number FIS12-037D
Price (SGD)
-
  • General imagepurpose scale measures radii from 1/16 to 3/8 in., angles in single degrees to 90°, .005 in. to .1mm scales, and line widths of .001, .002 and .003 in.
  • Inch Scale measures 3/4 in. in .005 in. intervals
  • Metric Scale measures 20mm in .1 mm intervals
  • Protractor Scale measures 360°degrees in single degrees
Specifications
For Use With (Application)To provide a flat image without distortion, measures radii from 0.06 in. to 0.37 in.
For Use With (Equipment)Lenscope™ magnifier
IncludesProtective case and storage pouch for case and lens
Product TypeGeneral Purpose Scale
Unit SizeEA