▸
Racks
▸
Freezer Storage Racks
▸
Refrigerator/Freezer Storage Racks
Actual product may vary
Refrigerator/Freezer Storage Racks
For sample storage in refrigerator or freezer.
Have Questions?
Catalog Number
Dimensions (L x W x H)
Positions
Quantity
FIS12-009-43A
9.5 x 9.5 x 2.5 in.
100
100
Catalog number
FIS12-009-43A
Price (SGD)
223.00
Each
Add to cart
Dimensions (L x W x H):
9.5 x 9.5 x 2.5 in.
Positions:
100
Quantity:
100
Price (SGD)
223.00
Each
Add to cart
For sample storage in refrigerator or freezer.
Clear acrylic racks store 16mm O.D. specimen tubes in refrigerator or freezer
Allow sufficient space between tubes to let you read individual tube labels
Make efficient use of shelf space
Handgrips for easy carrying
Specifications
Autoclavable
Not Autoclavable
Color
Transparent
Holds
Tubes (16 mm)
Material
Acrylic
Positions
100
Quantity
100
Dimensions (L x W x H)
9.5 x 9.5 x 2.5 in.
Length (Metric)
24.1 cm
Length (English)
9.5 in.
Width (English)
9.5 in.
Height (English)
2.5 in.
Product Type
Refrigerator/Freezer Rack
Diameter (Metric) Inner
16 mm
Height (Metric)
6.3 cm
Width (Metric)
24.1 cm