Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Clamps and Supports
▸
Stands and Rings
▸
Research Grade Retort Base and Rod Set
Actual product may vary
Eisco
Research Grade Retort Base and Rod Set
Available with various sized rods
Have Questions?
Change view
Catalog Number
Diameter (Metric) Rod
Length (Metric) Rod
Width (Metric)
FIS12-000-101
1 cm
49.5 cm
20.3 cm
FIS12-000-102
1 cm
60.9 cm
22.8 cm
FIS12-000-103
1.27 cm
90.1 cm
27.9 cm
3 Options
Catalog number
FIS12-000-101
Price (SGD)
83.00
Each
Add to cart
Diameter (Metric) Rod:
1 cm
Length (Metric) Rod:
49.5 cm
Width (Metric):
20.3 cm
Price (SGD)
83.00
Each
Add to cart
Has Cast iron base
Epoxy powder coated blue base
Nickel Plated Steel Rod
Specifications
Material
Cast Iron (Base), Epoxy Powder (Coating), Nickel Plated Steel (Rod)
Height (Metric)
5.08 cm
Length (English) Base
5 in.
Length (English) Rod
19.5 in.
Length (Metric) Base
12.7 cm
Length (Metric) Rod
49.5 cm
Width (English)
8 in.
Width (Metric)
20.3 cm
Diameter (English) Rod
0.4 in.
Diameter (Metric) Rod
1 cm
Includes
Steel Base, Steel Rod
Product Type
Retort Base w/Rod
Size
Medium