Magic Touch 2™ Ice Pans
Actual product may vary
Magic Touch 2™ Ice Pans
Magic Touch 2™ Ice Pans
Bel-Art
Magic Touch 2™ Ice Pans
Superior insulating properties combined with high chemical resistance
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)ColorIncludesLength (Metric)
FIS11-999-0511 LBlack EbonyPour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base18.3 cm
FKS134224391 LTrue BluePour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base18.3 cm
FKS134824394 LTrue BluePour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base30.4 cm
Catalog number FIS11-999-051
Price (SGD)
239.00
Each
Capacity (Metric):
1 L
Color:
Black Ebony
Includes:
Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base
Length (Metric):
18.3 cm
Price (SGD)
239.00
Each
Superior insulating properties combined with high chemical resistance
  • Compatible not only with wet ice, but also with ultra-low temperature materials including dry ice, salt slurries, dry ice solvent slurries, or liquid nitrogen
  • Able to withstand a wide temperature range of -196°C (-320°F) to 100°C (212°F)
  • High-quality, expanded urethane laboratory ice pans are lightweight, durable, non-sweating and impervious to moisture and odors
  • Lightly textured, non-slip finish and raised edge provides secure transport and a stylish look
  • Feature a convenient pour spout, drip-proof rims, a fill line indicator, and a snug-fitting, keyed lid and base for easy stacking
  • 1L pan does not include pour spout or fill line indicator
Specifications
Height (Metric)11.6 cm
Height (English)4.5 in.
Length (Metric)18.3 cm
Length (English)7.25 in.
Width (Metric)18.3 cm
Width (English)7.25 in.
Capacity (English)0.227 gal.
IncludesPour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base
DesignLightly textured, Non-slip
MaterialUrethane
TypeLaboratory Ice Pan
ColorBlack Ebony
Capacity (Metric)1 L
Holds24 Microtubes
AutoclavableNot Autoclavable
Temperature ControlNo
Temperature Range-196°C to +100°C
Unit SizeEach