E-Series Handheld Lamps
Actual product may vary
Spectroline
E-Series Handheld Lamps
A full range of models in this lamp series
Have Questions?
Catalog NumberLighting IntensityDimensions (L x W x H)
FIS11-992-23300/310μW/cm28 x 3.25 x 2.5 in. (203.2 x 82.55 x 63.5 mm)
Catalog number FIS11-992-23
Price (SGD)
1,367.00
Each
Lighting Intensity:
300/310μW/cm2
Dimensions (L x W x H):
8 x 3.25 x 2.5 in. (203.2 x 82.55 x 63.5 mm)
Price (SGD)
1,367.00
Each
Select the best combination of UV wavelength, intensity, size, and wattage for specific applications, including chromatography, gel electrophoresis, UV irradiation, and UV curing.
  • Offer easy wavelength interchangeability, making these viewing systems ideal for numerous applications where a darkened environment is needed
  • Specular aluminum reflector ensures optimum UV irradiance
  • Corrosion-resistant, specular aluminum housings provide comfortable, long-lasting service
  • All short- and medium-wave models feature Longlife™ filter glass for higher initial UV transmission and maximum resistance to solarizations
  • Outfitted with convenient, removable carrying handle for use with lamp stand
  • Available in 120V/60Hz, 230V/50Hz, or 100V/50-60Hz
Specifications
Voltage120 V
Wavelength365/254 nm
Case MaterialSilver Anodized Aluminum Housing
Dimensions (L x W x H)8 x 3.25 x 2.5 in. (203.2 x 82.55 x 63.5 mm)
Electrical Requirements120 V, 60 Hz
For Use With (Equipment)RUVIS
Height (English)2.5 in.
Height (Metric)63.5 mm
IncludesOne 4 W longwave tube, one 4 W shortwave tube with longlife filter assembly
Length (English)8 in.
Length (Metric)203.2 mm
Lighting Intensity300/310μW/cm2
Packaging Quantity1 Ea.
Width (English)3.25 in.
Width (Metric)82.55 mm
Product TypeHandheld UV Lamp
Frequency60 Hz
Unit SizeEach