UV Absorbing Goggles
Actual product may vary
Spectroline
UV Absorbing Goggles
Designed for long wearing comfort and protection against most ultraviolet light sources
Have Questions?
Catalog NumberProduct Type
FIS11-992-148Goggles
Catalog number FIS11-992-148
Price (SGD)
117.00
Each
Product Type:
Goggles
Price (SGD)
117.00
Each
  • Improve contrast between the fluorescent area and the background by eliminating the “blue haze” interference
  • Goggles (UVG–50) provide maximum protection from extended or high intensity UV exposure and adjust to fit the face
Specifications
Product TypeGoggles
Unit SizeEach