Kik-Step™ Stool
Actual product may vary
Cramer
Kik-Step™ Stool
The Kik-Step™ Stool is a rolling stool with a rubber bumper to protect walls and equipment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FIS11-931Gray
FIS11-931-005Silver
Catalog number FIS11-931
Price (SGD)
-
Color:
Gray
The Kik-Step™ Stool is a rolling stool with a rubber bumper to protect walls and equipment.
  • Step on it and spring-loaded casters retract for safe, sure footing
  • Step off and stool rolls smoothly
  • Two steel steps cushioned with rubber treads
  • Epoxy finish protected by wide rubber bumper around base
Specifications
ColorGray
Diameter (English) Base15.5 in.
Diameter (Metric) Base39.37 cm
FinishEpoxy
Height (English)14.5 in.
Height (Metric)36.83 cm
IncludesRubber treads and base bumper, retractable casters
Warranty10 Years
Weight (English)10 lb.
Weight (Metric)4.53 kg
Load Capacity (English)350 lb.
Load Capacity (Metric)158.75 kg
MaterialStainless Steel
Product TypeStep Stool
Unit SizeEach