Xtra™ Black Utility Carts
Actual product may vary
Xtra™ Black Utility Carts
Large shelves hold small baths, centrifuges, pH meters, and other equipment
Have Questions?
Catalog number FIS11-926-42
Price (SGD)
2,427.00
Each
Price (SGD)
2,427.00
Each
Black, four-shelf utility cart
  • Load capacity: 136.8kg (300 lb.)
  • Tough HDPE construction resists chips, dents, scratches, and rust
  • Easy-to-clean shelves with rounded interior corners and raised edges to keep small items from slipping off
  • Ergonomically designed handles on both cart ends
  • Quiet-rolling, nonmarking 10.2cm (4 in.) diia. casters
  • Choice of three-shelf models in three styles: open, enclosed on two ends, and enclosed on three sides (virtually a cabinet on wheels)
  • Also available in four-shelf open style
Specifications
Capacity (English) Shelf100 lb.
Capacity (Metric) Shelf45.4 kg
Product TypeInstrument Cart
ColorGray
Dimensions (L x W x H)40.62 x 20 x 37.8 in. (103.17 x 50.8 x 96.01 cm)
For Use With (Application)Securely transporting tools and smaller equipment in hospitals, healthcare centers and labs
Height (English)37.8 in.
Height (Metric)96.01 cm
IncludesLockable doors, enclosed end panels on 3 sides, 3 easy-to-clean shelves with rounded interior corners and raised edges, ergonomically handles, quiet-rolling nonmarking 4 in. dia. casters
Length (English)40.62 in.
Length (Metric)103.17 cm
Load Bearing Capacity (English)300 lb.
Load Bearing Capacity (Metric)136 kg
MaterialHDPE
No. of Shelves3
Width (English)20 in.
Width (Metric)50.8 cm
Unit SizeEach