Steri-Wrap™ I Sterilization Wrap
Actual product may vary
Steri-Wrap™ I Sterilization Wrap
Steri-Wrap™ I Sterilization Wrap
Steri-Wrap™ I Sterilization Wrap
Propper
Steri-Wrap™ I Sterilization Wrap
Steri-Wrap™ I is a cellulose based medium weight wrap, ideal for general hospital use in steam and ethylene oxide sterilization cycles. Efficient microbial barrier, high-level of protection against fluids and particles. Highly permeable meaning no wet packs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Width (Metric)
FIS11-890-8B38.1 cm38.1 cm
FIS11-890-8A30.4 cm30.4 cm
FIS11-890-8C50.8 cm50.8 cm
Catalog number FIS11-890-8B
Price (SGD)
861.00
Case
Length (Metric):
38.1 cm
Width (Metric):
38.1 cm
Price (SGD)
861.00
Case
  • Steri-Wrap™ is cellulose based sterilization wrap available in three grades for keeping packages safe and secure during the sterilization process.
  • Steri-Wrap™ I is a medium weight wrap, ideal for general hospital use.
  • Steri-Wrap™ is designed to be virtually lint free with tightly anchored fibers.
  • Wrap is reversible and features excellent liquid repellency.
  • Steri-Wrap™ can be used in steam or ethylene oxide sterilization.
  • Variety of of fabric strengths to meet your needs.
Specifications
AutoclavableAutoclavable
ColorBlue
DisposableSingle-use
For Use With (Application)General hospital use
MaterialNon-woven Fabric
SterilityNon-sterile
Dimensions (L x W)15 x 15 in. (38.1 x 38.1 cm)
Length (English)15 in.
Width (English)15 in.
Product TypeSteri-Wrap I
Length (Metric)38.1 cm
Width (Metric)38.1 cm
Unit SizeCase