Search Fisher Scientific
| Catalog Number | Color | Dimensions (L x W) | Diameter (English) |
|---|---|---|---|
| FIS11-865-109 | White, Yellow, Green | 13.7 m x 1.9 cm | 1 in. |
| FIS03-411-621 | Blue | 36.4 m x 12.7 mm | 3 in. |
| FIS03-411-607 | Green | 13.7 m x 19 mm | 1 in. |
| FIS03-411-622 | Blue | 36.4 m x 19 mm | 3 in. |
| FIS03-411-616 | Yellow | 36.4 m x 19 mm | 3 in. |
| FIS03-411-601 | White | 13.7 m x 19 mm | 1 in. |
| FIS03-411-619 | Green | 36.4 m x 19 mm | 3 in. |
| FIS03-411-600 | White | 13.7 m x 12.7 mm | 1 in. |
| FIS03-411-628 | White | 54.6 m x 19 mm | 3 in. |
| FIS03-411-625 | Orange | 36.4 m x 19 mm | 3 in. |
| FIS03-411-610 | White | 36.4 m x 19 mm | 3 in. |
| FIS03-411-604 | Yellow | 13.7 m x 19 mm | 1 in. |
| FIS03-411-613 | Red | 36.4 m x 19 mm | 3 in. |
| FKS13434720 | Yellow | 36.4 m x 12.7 mm | 3 in. |
| FKS13455246 | Yellow | 36.4 m x 25.4 mm | 3 in. |