ThermoSafe Insulated Storage and Transport Chests
ThermoSafe Insulated Storage and Transport Chests
Actual product may vary
ThermoSafe Insulated Storage and Transport Chests
Dry ice storage chests provide mobile, non-electrical low-temperature maintenance for a wide variety of laboratory and field applications. Ideal for dry ice or wet ice storage, hot storage, or field collection.
Have Questions?
Catalog NumberDimensions (L x W x H)TypeDesignClosure Type
FIS11-676-1123 x 22 x 20.75 in.Storage and Transport Chest2 Spring Loaded HandlesHinged
Catalog number FIS11-676-11
Price (SGD)
2,862.00
EA
Add to cart
Dimensions (L x W x H):
23 x 22 x 20.75 in.
Type:
Storage and Transport Chest
Design:
2 Spring Loaded Handles
Closure Type:
Hinged
Price (SGD)
2,862.00
EA
Add to cart
Ideal for dry ice and wet ice storage, short-term frozen and hot storage.
  • Hard cased units that provide low-cost storage. Units are low maintenance and easy to clean.
  • Available in different sizes for different durations.
  • Dollies are sold separately.
  • Long term usage, all components can be replaceable.
  • Units must be filled, or dunnage added to the unit, for maximum average holding time.
Specifications
Height (Metric)52.70 cm
Height (English)20.75 in.
Length (Metric)58.42 cm
Length (English)23 in.
Width (Metric)55.88 cm
Width (English)22 in.
Dimensions (L x W x H)23 x 22 x 20.75 in.
Closure TypeHinged
For Use With (Equipment)Sonoco™ ThermoSafe Aluminum Dollies (Fisher Cat. 11-676-24)
For Use With (Application)Ideal for dry ice and wet ice storage, short-term frozen and hot storage
Weight (English)35 lb.
Design2 Spring Loaded Handles
MaterialPolyethylene
TypeStorage and Transport Chest
ColorGray
Capacity (Metric)70.4 L
Unit SizeEA