Magic Touch™ Insulated Lab Pans
Magic Touch™ Insulated Lab Pans
Actual product may vary
Bel-Art
Magic Touch™ Insulated Lab Pans
Keep temperature sensitive samples in an optimal environment. Bel Art™ SP Scienceware™ Magic Touch™ Insulated Lab Pans are the lightest icewares on the market.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)ColorMaterial
FIS11-675-1239 LBluePolymer Foam
FIS11-676-38A4 LBluePolymer Foam
FIS11-675-589 LBlackPolymer Foam
Catalog number FIS11-675-123
Price (SGD)
-
Capacity (Metric):
9 L
Color:
Blue
Material:
Polymer Foam
Keep temperature sensitive samples in an optimal environment. Bel Art™ SP Scienceware™ Magic Touch™ Insulated Lab Pans are the lightest icewares on the market.
  • Large enough to accommodate circular and rectangular floating test tube racks, flasks, bottles, etc.
  • Expanded polyvinyl chloride, therefore unbreakable under normal conditions
  • Withstand temperatures from -196° to +93°C (-321° to +200°F) without sweating or leaking
  • Will not absorb odors or moisture
  • Chemically resistant to highly acidic and basic solutions
  • Suitable for use with ice, dry ice alcohol slurries, or liquid nitrogen
  • Stackable
  • Wall thickness: 0.75 in. (2cm)
Specifications
Height (Metric)13 cm
Height (English)5.1 in.
Length (Metric)41 cm
Length (English)16 in.
Width (Metric)33 cm
Width (English)13 in.
Dimensions (L x W x H)16 x 13 x 5.1 in. (41 x 33 x 13 cm)
For Use With (Application)Ideal for storing and transporting temperature sensitive samples in Ice, Alcohol Slurries, Water baths and more
MaterialPolymer Foam
TypeMaxi Lab Ice Pan
ColorBlue
Capacity (Metric)9 L
Temperature Range-196°C to +93°C
Unit SizeEA