5-Hydroxyindole, 98+%
5-Hydroxyindole, 98+%
5-Hydroxyindole, 98+%
Tempshield
5-Hydroxyindole, 98+%
CAS: 1953-54-4 | C8H7NO | 133.15 g/mol
Have Questions?
Catalog NumberQuantity
FIS11-394-3241 g
Catalog number FIS11-394-324
Price (SGD)
517.00
Pair
Quantity:
1 g
Request bulk or custom format
Price (SGD)
517.00
Pair
Specifications
Chemical Name or Material5-Hydroxyindole
CAS1953-54-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c