Potash, sulfurated, S 11.8% min as sulfide
Potash, sulfurated, S 11.8% min as sulfide
Potash, sulfurated, S 11.8% min as sulfide
Tempshield
Potash, sulfurated, S 11.8% min as sulfide
Mixture composed of potassium polysulfides and potassium thiosulfate | CAS: 39365-88-3 | K4O3S3 | 300.57 g/mol
Have Questions?
Catalog NumberQuantity
FIS11-394-308100 g
Catalog number FIS11-394-308
Price (SGD)
742.00
Pair
Quantity:
100 g
Request bulk or custom format
Price (SGD)
742.00
Pair
Specifications
Chemical Name or MaterialPotash
Name NoteMixture composed of potassium polysulfides and potassium thiosulfate
CAS39365-88-3
ColorGreen
Health Hazard 1H228-H314-H335
View more
Potash is used in black/brown patina (copper, brass, bronze). Its use include decorative color effects on copper, brass, bronze and nickel. Sulfurated potash has been used in the treatment of acne and other skin disorders usually in the form of a lotion with zinc sulfate.