Hot-Hand™ Silicone Rubber Grip Mitts
Actual product may vary
Bel-Art
Hot-Hand™ Silicone Rubber Grip Mitts
Mitts feature concave tipped studs for a non-slip grip, and are capable of withstanding temperatures up to 500°F.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FIS11-394-300Red
FIS08-647-729Blue
Catalog number FIS11-394-300
Price (SGD)
85.00
Each
Color:
Red
Price (SGD)
85.00
Each
Mitts feature concave tipped studs for a non-slip grip, and are capable of withstanding temperatures up to 500°F.
  • Protection when handling hot or cold objects, including dry ice
  • Capable of withstanding temperatures as high as 260°C (500°F)
  • Will remains flexible at temperatures as low as -57°C (-70°F)
  • Silicone rubber naturally conforms to hand and object
  • Concave tipped studs provide a non-slip grip
  • Easier and faster to use than gloves or tongs for many applications
  • One size fits all hands, left or right
Specifications
Accelerator FreeNo
ApplicationsLaboratory
Coating MaterialUncoated
ColorRed
HandAmbidextrous
Latex FreeYes
MaterialSilicone Rubber
Product TypeHand/Finger Protector
SizeUniversal
StyleFull-hand Mitt
Temperature (English) Max.500°F
Temperature (Metric) Max260°C
For Use With (Application)Handling Hot/Cold Objects
SeriesHot Hand™
Temperature (English) Min-70°F
Temperature (Metric) Min.-57°C
Unit SizeEach