AlphaTec™ Solvex™ 37-145 Nitrile Gloves
Actual product may vary
AlphaTec™ Solvex™ 37-145 Nitrile Gloves
AlphaTec™ Solvex™ 37-145 Nitrile Gloves
AlphaTec™ Solvex™ 37-145 Nitrile Gloves
Ansell
AlphaTec™ Solvex™ 37-145 Nitrile Gloves
Solvex™ 11 mil nitrile gloves are tactile and durable and can be used in a diverse range of applications including automotive, machinery/equipment and chemical.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS11-391-136D9
FIS11-391-136B7
FIS11-391-136C8
Catalog number FIS11-391-136D
Price (SGD)
92.00
Pack
Size:
9
Price (SGD)
92.00
Pack
Solvex™ 11 mil nitrile gloves are tactile and durable and can be used in a diverse range of applications including automotive, machinery/equipment and chemical.
  • Nitrile construction protects against abrasions, cuts, snags and punctures
  • Curved finger design and contoured palm help reduce fatigue
  • Full cut for easy donning and doffing
  • Dark Green
  • No inner cotton flocking, reducing risk of lint contamination
  • Sandpatch finish provides smooth surface
  • Fully reusable, comfortable
  • Complies with FDA food handling requirements
Specifications
ANSI Abrasion RatingANSI 5
Certifications/ComplianceFDA 21 CFR 177.2600
ChlorinatedYes
ColorGreen
Cut Resistance RatingANSI A4
Latex FreeYes
MaterialNitrile
Size9
Thickness (English)11 mil
Thickness (Metric)0.27 mm
EN388 StandardEN 388:4001, EN 388:3001X
EN 421No
EN ISO 374-1Type A
EN ISO 374-1 CHEMICAL LETTER CODESJKNOPT
EN ISO 374 - 5Bacteria, Fungi only
For Use With (Application)Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Glove Cuff StyleGauntlet
Grip DesignSandpatch
LiningUnlined
Model37-145 9 117075
Length (English)13 in.
Length (Metric)33 cm
Unit SizePack