Fiberboard Cryogenic Storage Boxes and Dividers
Actual product may vary
Fiberboard Cryogenic Storage Boxes and Dividers
Create customized storage box configurations
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H)
FIS10-987-06713.3 x 13.3 x 2.5 m (5.25 x 5.25 x 1 in.)
FIS10-987-0657.6 x 7.6 x 2.5 cm (3 x 3 x 1 in.)
FIS03-391-5147.6 x 7.6 x 5 cm (3 x 3 x 2 in.)
Catalog number FIS10-987-067
Price (SGD)
17.00
Each
Dimensions (L x W x H):
13.3 x 13.3 x 2.5 m (5.25 x 5.25 x 1 in.)
Price (SGD)
17.00
Each
  • Snug-fitting lid prevents accidental separation if dropped
  • Use in mechanical freezers or liquid nitrogen tanks to –196°C
  • Constructed of sturdy cardboard with moisture-repellent coating
  • Designed with small holes on the bottom to allow for drainage
These economical cryo/freezer storage boxes offer excellent low-temperature storage. Moisture-repellent coating protects boxes from effects of liquid nitrogen or condensation. Choose from a wide range of boxes and dividers to accommodate tubes from 0.5 to 50 mL. Boxes can be ordered with dividers for a complete box configuration or without dividers to customize your storage needs. The wide range of dividers can hold from 16 to 196 tubes with diameters from 8 to 33.5 mm. Order additional and replacement cell dividers separately.
Specifications
ColorWhite
MaterialCardboard
ShapeSquare
TypeFreezer Box
For Use With (Application)Holding Single PCR Tubes or Strips of 8
Holds196 Tubes
Tube Array14 x 14
Dimensions (L x W x H)13.3 x 13.3 x 2.5 m (5.25 x 5.25 x 1 in.)
Unit SizeEach