Ceramic Shelves for Isotemp Furnaces
Ceramic Shelves for Isotemp Furnaces
Actual product may vary
Ceramic Shelves for Isotemp Furnaces
This Accessory piece is a freestanding shelf with legs. Surfaces on top and underneath can both be utilized.
Have Questions?
Catalog NumberStyle
FIS10-651SNFreestanding
Catalog number FIS10-651SN
Price (SGD)
-
Style:
Freestanding
This Accessory piece is a freestanding shelf with legs. Surfaces on top and underneath can both be utilized.
Accessory freestanding shelf with legs for 0.58 and 1.26 cu. ft. furnaces. Use surface on top and underneath.
  • 0.58 cu. ft. furnace takes one shelf
  • 1.26 cu. ft. furnace takes two shelves
  • Replacement flat shelf is for 1.26 cu. ft. furnaces
  • All shelves support 3 lb. (1.4kg)
Specifications
For Use With (Equipment)0.58 and 1.26 cu. ft. isotemp furnaces
TypeShelf
MaterialCeramic
StyleFreestanding
Unit SizeEA