Safety Blades
Safety Blades
Actual product may vary
Safety Blades
Safety Blades
Thermo Scientific
Safety Blades
Finger-friendly™ blade is safer to touch than traditional blades
Have Questions?
Catalog NumberLength (English)Style
FIS10-5282.56 in.Pointed Tip
Catalog number FIS10-528
Price (SGD)
426.00
CS
Add to cart
Length (English):
2.56 in.
Style:
Pointed Tip
Price (SGD)
426.00
CS
Add to cart
  • Finger-friendly™ edge stays sharp 11x longer
  • Reduces injuries, lowers costs
  • Non-sparking, non-conductive, non-magnetic
  • Safe up to 1600 degrees Celsius
  • Chemically inert, never rusts
  • No-tool blade change
  • Fewer blade changes = fewer injuries
Specifications
Product TypeSafety Blade
For Use With (Equipment)Sice™ handles: 10550, 10554, 10558, 10585
Length (English)2.56 in.
Length (Metric)65 mm
MaterialZirconium Oxide
ColorWhite
For Use With (Application)Safety Cutting
Includes3 dual-sided Utility Knife blades
No. per Pack3 Pk.
StylePointed Tip
Weight (English)0.17 oz.
Weight (Metric)4.8 g
Width (English)0.47 in.
Width (Metric)12 mm
Unit SizeCS