Heavy-Duty/Utility Funnels
Heavy-Duty/Utility Funnels
Actual product may vary
Heavy-Duty/Utility Funnels
Heavy-Duty/Utility Funnels
Heavy-Duty/Utility Funnels
Ideal for continuous lab work and heavy-duty jobs
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric)TopHeight (Metric)
FIS10-500-540 mL6.5cm9 cm
FIS10-500-10500 mL20.3 cm20.5 cm
Catalog number FIS10-500-5
Price (SGD)
80.00
Pack
Add to cart
Capacity (Metric):
40 mL
Diameter (Metric)Top:
6.5cm
Height (Metric):
9 cm
Price (SGD)
80.00
Pack
Add to cart
Ideal for continuous lab work and heavy-duty jobs
  • Durable polypropylene construction with external ridges and ribbing to prevent vapor lock
  • Autoclavable to 120°C (248°F)
Specifications
Diameter (Metric)Top6.5cm
Capacity (Metric)40 mL
MaterialPolypropylene
Height (Metric)9 cm