▸
Secondary Antibodies
▸
Goat Secondary Antibodies
▸
Kappa Goat anti-Mouse, TRITC (Rhodamine), Polyclonal, Southern Biotech™
Actual product may vary
Kappa Goat anti-Mouse, TRITC (Rhodamine), Polyclonal, Southern Biotech™
Goat Polyclonal Secondary Antibody
Have Questions?
Catalog Number
Quantity
FIS10-500-3
1.0 mg
Catalog number
FIS10-500-3
Price (SGD)
59.00
Pack
Add to cart
Quantity:
1.0 mg
Price (SGD)
59.00
Pack
Add to cart
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
Antigen
Kappa
Applications
ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification
Polyclonal
Concentration
1.0mg/mL
Conjugate
TRITC
Formulation
Supplied in PBS/NaN3
Host Species
Goat
Immunogen
Mouse κ light chains
Purification Method
Affinity Purified
Quantity
1.0 mg
Regulatory Status
RUO
Primary or Secondary
Secondary
Target Species
Mouse
Isotype
IgG
Unit Size
Pack
Contents & Storage
Store at 2-8°C; Protect from light