Check Order Status
PYREX™ Plain Diameter 60° Angle Funnel with Short Wide Stem
Actual product may vary
PYREX™ Plain Diameter 60° Angle Funnel with Short Wide Stem
Beaded rims reduce chipping
Have Questions?
Change view
Catalog Number
Diameter (Metric)Top
Height (Metric)
FIS10-373B
14.7 cm
22.8 cm
FIS10-373A
12.2 cm
19.9 cm
2 Options
Catalog number
FIS10-373B
Price (SGD)
1,047.00
PK
Add to cart
Diameter (Metric)Top:
14.7 cm
Height (Metric):
22.8 cm
Price (SGD)
1,047.00
PK
Add to cart
Beaded rims reduce chipping
Stems are approximately 15 mm O.D. by 100 mm (4) long.
Specifications
Diameter (Metric)Top
14.7 cm
Diameter (Metric) Stem
1.5 cm
Material
Borosilicate Glass
Height (Metric)
22.8 cm
Angle
60°
Length (Metric) Stem
10 cm
Product Type
Plain Filling Funnel