Nalgene™ Büchner Two-Piece Polypropylene Funnels
Actual product may vary
Nalgene™ Büchner Two-Piece Polypropylene Funnels
Nalgene™ Büchner Two-Piece Polypropylene Funnels
Nalgene™ Büchner Two-Piece Polypropylene Funnels
Nalgene™ Büchner Two-Piece Polypropylene Funnels
Nalgene™ Büchner Two-Piece Polypropylene Funnels
Thermo Scientific
Nalgene™ Büchner Two-Piece Polypropylene Funnels
Separate the lightweight and durable two-piece polypropylene funnels for easy cleaning. These funnels won't chip, crack or break.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric)Top
FIS10-362G2.91 L175 mm
FIS10-362C186 mL75 mm
4280-0900358 mL99 mm
Catalog number FIS10-362G
Price (SGD)
210.00
Each
Capacity (Metric):
2.91 L
Diameter (Metric)Top:
175 mm
Price (SGD)
210.00
Each
Separate the lightweight and durable Thermo Scientific™ Nalgene™ Büchner Two-Piece Polypropylene Funnels for easy cleaning. These autoclavable funnels won't chip, crack or break making them a valuble asset to lab safety programs.
  • Sturdy funnels consist of cup and funnel with perforated plate for easy cleaning and good chemical resistance
  • All sizes are made entirely of polypropylene and are autoclavable to 135°C (275°F)
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications
Diameter (Metric)Top175 mm
Capacity (Metric)2.91 L
MaterialPolypropylene
Height (Metric)285 mm
AutoclavableAutoclavable
DescriptionFunnel Buchner PP 150mm
Length (Metric) Stem107mm
Quantity1/Pk.; 4/Cs.
Unit SizeEach