PYREX™ Plain 60° Angle Funnel with Long Narrow Stem
PYREX™ Plain 60° Angle Funnel with Long Narrow Stem
Actual product may vary
PYREX™ Plain 60° Angle Funnel with Long Narrow Stem
Permit precise filter paper fitting
Have Questions?
Catalog NumberDiameter (Metric)TopDiameter (Metric) StemHeight (Metric)
FIS10-326D10 cm0.9 cm23.6 cm
Catalog number FIS10-326D
Price (SGD)
351.00
CS
Add to cart
Diameter (Metric)Top:
10 cm
Diameter (Metric) Stem:
0.9 cm
Height (Metric):
23.6 cm
Price (SGD)
351.00
CS
Add to cart
These funnels have a beaded edge for strength. They are fabricated from blanks formed to a 60° angle, which permits accurate fitting of the filter paper, thus reducing filtering time. The stems are made of heavy wall tubing approximately 150 mm (6) long.
Specifications
Diameter (Metric)Top10 cm
Diameter (Metric) Stem0.9 cm
MaterialBorosilicate Glass
Height (Metric)23.6 cm
Angle60°
Length (Metric) Stem15 cm
Product TypePlain Filling Funnel
Unit SizeCS