General-Purpose Pinning Forceps
General-Purpose Pinning Forceps
Actual product may vary
General-Purpose Pinning Forceps
General-Purpose Pinning Forceps
General-Purpose Pinning Forceps
Fisherbrand General-Purpose Pinning Forceps makes pinning zoological specimens easy, even when space is limited.
Have Questions?
Catalog NumberLength (Metric)
FIS10-27011.4 cm
Catalog number FIS10-270
Price (SGD)
17.00
EA
Add to cart
Length (Metric):
11.4 cm
Price (SGD)
17.00
EA
Add to cart
Our instruments are designed to exceed performance expectations -This is the Fisherbrand Advantage.
  • Make pinning small zoological specimens easy
  • Angled, Blunt, Serrated Tips
  • Grooved handles for better grip
  • Made from corrosion-resistant ASTM™ standard stainless steel
  • Electronically passivated for corrosion resistance
  • Rockwell hardness (HRC): 40--45
  • Latex-free
  • Can be sterilized by Eto/Gamma radiation; Fully Autoclavable
  • Quality control per U.S. Government specs
  • Produced under ISO 9002, GMP, cGMP guidelines
Specifications
Tip StyleCurved
Length (English)4.5 in.
MaterialStainless Steel
Length (Metric)11.4 cm
Product TypeGeneral Purpose Forceps
Quantity (No. per Case)12 Case
Unit SizeEA