PYREXPLUS™ Coated Class A Volumetric Flask with Glass Standard Taper Stopper
PYREXPLUS™ Coated Class A Volumetric Flask with Glass Standard Taper Stopper
Actual product may vary
PYREXPLUS™ Coated Class A Volumetric Flask with Glass Standard Taper Stopper
PVC coating on the outside reduces chance of breakage
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS10-240H2000 mL
FIS10-240G1 L
Catalog number FIS10-240H
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
2000 mL
Flask features a protective PVC coating for longer product life and safety. Protective coating helps prevent glass from shattering and reduces spills. Autoclavable (121°C) and resistant to thermal shock. The graduation mark is easy to read. Supplied with standard taper PYREX stopper.
For stopper only, see Cat. No. 7650.
Warning: Do not place over direct heat or flame. Do not heat above 121°C moist heat or 110°C dry heat.
Specifications
Closure MaterialGlass
Diameter (English) Outer6.22 in.
Capacity (Metric)2000 mL
MaterialPyrexplus
Height (English)14.05 in.
Height (Metric)35.7 cm
Neck StyleLong
No. per Case2/Cs.
No. per Pack1/Pk.
Surface CoatingPyrexplus
Tolerance±0.5 mL
ClassA
Bottom ShapeFlat
Unit SizeEA