Kimble™ KIMAX™ Flasks Serialized and Certified with standard taper Pennyhead Stopper
Kimble™ KIMAX™ Flasks Serialized and Certified with standard taper Pennyhead Stopper
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Flasks Serialized and Certified with standard taper Pennyhead Stopper
Calibrated and certified against NIST-Traceable equipment
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS10-212B100 mL
FIS10-212A50 mL
Catalog number FIS10-212B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
100 mL
Calibrated and certified against NIST-Traceable equipment
  • Permanent individual serial no
  • Calibrated and certified against NIST-Traceable equipment
Specifications
Closure MaterialGlass
Capacity (Metric)100 mL
MaterialBorosilicate Glass
Height (English)7.08 in.
Height (Metric)18 cm
Tolerance±0.08 mL
Closure Size13
Unit SizeEA