PYREX™ Class A Volumetric Flasks with Glass Standard Taper Stopper
PYREX™ Class A Volumetric Flasks with Glass Standard Taper Stopper
Actual product may vary
PYREX™ Class A Volumetric Flasks with Glass Standard Taper Stopper
Machine-blown body
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeBottom ShapeTolerance
FIS10-210K4 L38Flat±1 mL
FIS10-210C100 mL13Flat±0.01 mL
FIS10-210G1 L22Flat±0.3 mL
Catalog number FIS10-210K
Price (SGD)
1,595.00
CS
Add to cart
Capacity (Metric):
4 L
Closure Size:
38
Bottom Shape:
Flat
Tolerance:
±1 mL
Price (SGD)
1,595.00
CS
Add to cart
Machine-blown body
The strength of these flasks has been increased appreciably through machine-blown bodies to which are sealed heavy-tubing necks tooled for standard taper glass stoppers. The graduation line is sharp and permanent; large white block letters make the inscription easy to read. The 1 mL and 2 mL sizes are test tube shaped. Sizes 3L through 6L have pennyhead standard taper stoppers.
For stopper only, see Cat. Nos. 7650 and 7661.
Specifications
Closure MaterialGlass
Diameter (English) Outer8.07 in.
Capacity (Metric)4 L
MaterialBorosilicate Glass
Height (English)18.9 in.
Height (Metric)48 cm
Neck StyleLong
No. per Case1/Cs.
No. per Pack1/Pk.
Tolerance±1 mL
GraduatedYes
ClassA
Bottom ShapeFlat
Closure TypePenny Stopper
Diameter (Metric) Outer20.5 cm
Closure Size38
Unit SizeCS