Check Order Status
White Line Flasks with Standard Taper Stoppers
Actual product may vary
White Line Flasks with Standard Taper Stoppers
Single graduation ring blasted on the neck.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FIS10-205J
150 mL
FIS10-205H
20 mL
2 Options
Catalog number
FIS10-205J
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
150 mL
Calibrated “To Contain”
Reusable
Class A volumetric
Specifications
Calibration
Calibrated To Contain
Closure Material
Glass
Capacity (Metric)
150 mL
Material
Borosilicate Glass
No. per Case
6
Graduated
Yes
Closure Size
13