PYREX™ Class A Volumetric Flask with Polyethylene Snap-Cap
PYREX™ Class A Volumetric Flask with Polyethylene Snap-Cap
Actual product may vary
PYREX™ Class A Volumetric Flask with Polyethylene Snap-Cap
Calibrated To Contain
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS10-202F1 L
FIS10-202A50 mL
Catalog number FIS10-202F
Price (SGD)
194.00
EA
Add to cart
Capacity (Metric):
1 L
Price (SGD)
194.00
EA
Add to cart
The strength of these flasks has been increased appreciably through machine-blown bodies to which are sealed heavy-beaded, heavy-tubing necks, tooled for snap cap fit. The graduation line is sharp and permanent; large white block letters make the inscription easy to read. The 1 mL and 2 mL sizes are test tube shaped. The 10 mL through 2000 mL sizes are supplied with snap caps.
Cat. Nos. 5580-1 and 5580-2 have no snap caps for 1 mL and 2 mL sizes.
For snap caps only, see Cat. No. 7666.
Specifications
Certifications/ComplianceASTM E694, ASTM E542, ASTM E288
Closure ColorBlue
Closure MaterialHDPE
DescriptionTop style: Heavy duty rim
Diameter (English) Outer4.92 in.
Capacity (Metric)1 L
MaterialBorosilicate Glass
Height (English)12.2 in.
Height (Metric)31 cm
Neck StyleLong
No. per Case6/Cs.
No. per Pack1/Pk.
Tolerance±0.3 mL
GraduatedYes
ClassA
Bottom ShapeFlat
Closure TypeSnap Cap
Diameter (Metric) Outer125 mm
Unit SizeEA