Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Closure Size |
|---|---|---|
| FIS10-200E | 250 mL | 16 |
| FISFB40110 | 10 mL | 2 |
| FISFB4012000 | 2000 mL | 8 |
| FIS10-200B | 50 mL | 16 |
| FIS10-200D | 200 mL | 16 |
| FIS10-200C | 100 mL | 16 |
| FIS10-200F | 500 mL | 16 |
| FIS10-200G | 1000 mL | 16 |