Reusable Glass Class A Volumetric Flasks with Snap Caps
Reusable Glass Class A Volumetric Flasks with Snap Caps
Actual product may vary
Reusable Glass Class A Volumetric Flasks with Snap Caps
Class A accuracy–without a serial number
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FIS10-200B50 mL16
FISFB4011010 mL2
FISFB40120002000 mL8
FIS10-200D200 mL16
FIS10-200C100 mL16
FIS10-200E250 mL16
FIS10-200F500 mL16
FIS10-200G1000 mL16
Catalog number FIS10-200B
Price (SGD)
58.00
Each
Add to cart
Capacity (Metric):
50 mL
Closure Size:
16
Price (SGD)
58.00
Each
Add to cart
Class A accuracy–without a serial number
  • Calibrated “To Contain”
Specifications
CalibrationCalibrated To Contain
Certifications/ComplianceClass A, ASTM E288
Closure MaterialPolyethylene
Capacity (Metric)50 mL
MaterialGlass
No. per Case1
GraduatedYes
Closure Size16
Unit SizeEach