Cylindrical Dewar Flasks with Plastic Mesh Casing
Cylindrical Dewar Flasks with Plastic Mesh Casing
Actual product may vary
Pope Scientific
Cylindrical Dewar Flasks with Plastic Mesh Casing
Pope Scientific Cylindrical Dewar Flasks with Plastic Mesh Casing, Depth Inside: 11.88 in., 30.2cm, Diameter Outer: 3.38 in., 8.6cm, Diameter Inner: 2.75 in., 7cm, Height: 13.26 in., 33.7cm, Capacity: 1L, Cap Lid Type: Plastic Mesh Cover, For Use With: Liquid nitrogen service, Borosilicate Glass
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDepth (English) InsideDiameter (English) OuterDiameter (English) InnerHeight (English)
FIS10-197B11.88 in.3.38 in.2.75 in.13.26 in.
FIS10-197D12 in.7 in.5.62 in.14.37 in.
Catalog number FIS10-197B
Price (SGD)
-
Depth (English) Inside:
11.88 in.
Diameter (English) Outer:
3.38 in.
Diameter (English) Inner:
2.75 in.
Height (English):
13.26 in.
Pope Scientific Cylindrical Dewar Flasks with Plastic Mesh Casing, Depth Inside: 11.88 in., 30.2cm, Diameter Outer: 3.38 in., 8.6cm, Diameter Inner: 2.75 in., 7cm, Height: 13.26 in., 33.7cm, Capacity: 1L, Cap Lid Type: Plastic Mesh Cover, For Use With: Liquid nitrogen service, Borosilicate Glass
Evacuated to 10-5 torr or better and covered by a protective mesh
  • Fully silvered double-walled borosilicate glass
  • Metal casing surrounds only bottom portion of flask
  • Protective plastic mesh covers remaining outer surface of glass
  • Special vented polyethylene stopper minimizes evaporation
  • Weighted bases for stability
  • No pads between walls on cylindrical Dewars
Specifications
Cap Lid TypePlastic Mesh Cover
Depth (English) Inside11.88 in.
Depth (Metric) Inside30.2cm
Diameter (English) Outer3.38 in.
Diameter (Metric) Outer8.6cm
For Use With (Application)Liquid nitrogen service
Height (English)13.26 in.
Height (Metric)33.7cm
ModelLab Grade
Capacity (Metric)1 L
Depth (Metric) Interior30.2 cm
Diameter (English) Inner2.75 in.
Diameter (Metric) Inner7 cm
MaterialBorosilicate Glass
StyleCylindrical, Wide Mouth
Unit SizeEA